Learn More
Bullard™ EVA PAPR System with Double Bib Tychem QC Hoods
Evolutionary Air (EVA) Powered Air-Purifying Respirator (PAPR) use a battery-powered fan to draw in ambient air through a filter cartridge then delivers filtered air to the user's double bib hood via a breathing tube.
Supplier: Bullard™ EVA20TICT
Description
- Uniquely designed, streamlined form makes PAPR lightweight, eliminates bulky corners that can dig into the wearer's back, and balances the unit's weight for maximum comfort
- Easy-to-read fuel gauge displays remaining battery capacity at a glance
- Battery status can be checked at any time during use
- Bullard Active Flow Technology automatically responds to a worker's need for more or less air flow
- Two distinct audible alarms notify wearers of low battery and low air flow
- Alarms are designed so the sound of the alarm travels through the breathing tube directly to the wearer's ear
- Powered by a long-lasting 10,000-hour brushless motor
- Single filter design for easy use
- Filter locking tab notifies the user that the filter is in place with both a visual indication and audible click
Order Info
A NIOSH approved EVA PAPR system consists of the following properly-assembled components: (1) hood or mask; (2) breathing tube; (3) blower assembly; (4) purifying filter cartridge. Check model system components carefully before ordering.
Compliance
NIOSH approved.
Specifications
| 7 to 8 hours | |
| Double bib hood, ratchet suspension, quick-attach | |
| Acetate |
| Foam | |
| PAPR system, EVA blower, battery, charger, breathing tube | |
| EVA20TICT |