Learn More
METTLER TOLEDO EVA Karl Fischer Titrators
Catalog No. 014388509
Change view
Click to view available options
Model:
EVA V1 Titrator
EVA V3 Titrator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
014388509 EVA V3 Titrator
014388539 EVA V1 Titrator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 014388509 Supplier METTLER TOLEDO Supplier No. 30869282
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The EVA V1 and V3 volumetric Karl Fischer (KF) titrators are ideal for efficiently determining the water content of simple liquids, soluble solids, and more challenging insoluble samples.
  • High reliability and efficiency via the FFA™ control algorithm, a fast-response digital sensor, and state-of- the-art dispensing technology
  • 120,000 step dispensing resolution
  • Intuitive OneClick™ user interface simplifies operations and provides customizable shortcuts
  • Automated solvent exchange minimizes exposure to hazardous chemicals
  • Seamless LabX™ integration enhances data management and compliance with regulatory requirements
  • Modular design to meet versatile laboratory needs
  • The EVA KF Titrators are designed for efficient and accurate titration across various applications, including chemistry, food and beverage, and pharmaceuticals.
  • Measure water content in a wide range of samples, with a limit of quantification as low as 0.2 mg of water per sample.
  • Featuring the new FFA™ control algorithm and biamperometric indication, the EVA KF titrators deliver fast and accurate water content results for any application.
  • The detachable user-friendly touchscreen with OneClick™ interface streamlines workflows and reduces user errors, while robust data management capabilities allow for easy storage and analysis of results.
  • Automatic sample detection and titrant dosing minimizes chemical exposure, ensuring a safer, cleaner laboratory.
  • With a compact design that fits well in any laboratory, the EVA KF titrators combine innovative technology and practicality, making them an essential for reliable titration.
Specifications
Connector Types USB-A, RJ45, 4-pin M8 socket, 8-pin M9 socket, 4-pin power mini-DIN socket
Drift 25 mL/min
IP Rating None
Languages 10 Languages
Material PBT, Stainless Steel
For Use With Liquid and Soluble Samples, and Challenging Insoluble Solid Samples via Homogenizers
Model EVA V3 Titrator
Type Volumetric Karl Fischer Titrator
Width (English) 7.6 in.
Depth (English) 7.1 in.