missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ EVA Healthcare PAPR Kit

Catalog No. 19021486
Change view
Click to view available options
Model:
EVAPAN1
EVAPAN2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
19021486 EVAPAN2
19021485 EVAPAN1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19021486 Supplier Bullard™ Supplier No. EVAPAN2

missing translation for 'validMainframeMsg'

Bullard™ EVA Healthcare PAPR Kit is a complete, ready-to-use PAPR system with HEPA Filter and large, loose-fitting facepiece

  • Complete, ready-to-use PAPR system with HEPA filter and 20LF2L (large) loose-fitting facepiece
  • Blood-borne pathogen protection you can trust - Loose-fitting facepiece made exclusively with Dupont™ Tychem™ 2000 material
  • Air delivered where you need it - Soft PlenumTM delivery system delivers air over top and into the breathing zone
  • Maximum visibility - Wide angle facepiece lens with unobstructed view
  • Loose-fitting facepiece designed for use with a stethoscope
  • Assigned protection factor (APF) 2.5X greater than an N95 mask
  • Improves breathing and overall comfort compared to an N95 mask
  • No fit test required
  • Two selectable flow rate options - 7 CFM and 8.5 CFM - offers more options and comfort to the user
  • Spash and respiratory protection in one easy-to-use system
  • ASTM-F1670 / F1671 protection against Blood and Blood-borne pathogens

Specifications

Battery Life 4 to 16 hours
Battery Type Lithium Polymer
Certifications/Compliance NIOSH 42 CFR Part 84 (TC-21 C-0836)
Certifications (EN) EN12941:1998/A2:2008, CE0194
Description 25 APF (Assigned Protection Factor)
Color Black, White
Filter Type HEPA
Product Type PAPR Healthcare Systems and Kits
Flow Rate 7.0 CFM/8.5 CFM
For Use With (Application) Healthcare, EMS, Emergency Responders, Dentists
Includes EVA Blower (EVA1), Loose-Fitting Facepiece (20 LF2 L), Breathing Tube (PA20 LFBT), Airflow Indicator (PA1 AFI), EVA Battery (EVABAT1), HEPA Filter
Model EVAPAN2
Quantity 1 Ea.
Suspension Type Built-in Soft Suspension
Show More Show Less