missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ EVA Foam Ice Pans and Buckets with Lids

Catalog No. p-7084082
Change view
Click to view available options
Capacity (Metric):
1 L
2.5 L
4 L
9 L
Color:
Blue
Green
Purple
Includes:
Bucket with Lid
Pan with Lid
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Color Includes
03395150 1 L Blue Pan with Lid
03395151 1 L Green Pan with Lid
03395152 1 L Purple Pan with Lid
03395153 4 L Blue Pan with Lid
03395154 4 L Green Pan with Lid
03395155 4 L Purple Pan with Lid
03395156 9 L Blue Pan with Lid
03395157 9 L Green Pan with Lid
03395158 9 L Purple Pan with Lid
03395159 2.5 L Blue Bucket with Lid
03395160 2.5 L Green Bucket with Lid
03395161 2.5 L Purple Bucket with Lid
03395162 4 L Blue Bucket with Lid
03395163 4 L Green Bucket with Lid
03395164 4 L Purple Bucket with Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 03395150
Only null left

Includes

removable lid.

Fisherbrand™EVA Foam Ice Pans and Buckets with Lids provide excellent insulation for any benchtop cryogenic application, without the mess!

  • Sweat-proof ethylene vinyl acetate will not leak
  • Compatible with ice, dry ice, liquid nitrogen, alcohol, and saline solutions
  • Bases and lids feature glove-friendly handles
  • Cleanup is quick and easy with molded pour spouts

Specifications

For Use With (Equipment) Ice, dry ice, liquid nitrogen, alcohol, and saline solutions
Includes Pan with Lid
Description Pan with Lid
Material EVA
Type Cryo-Safe Cold Box
Quantity 1
Color Blue
Capacity (Metric) 1 L
Autoclavable Not Autoclavable
Temperature Control No