missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Europium(III) chloride hexahydrate, 99.9%, (trace metal basis), -10 mesh
CAS: 13759-92-7 | Cl3Eu·6H2O | 366.4 g/mol
$264.30 - $264.30
Chemical Identifiers
| CAS | 13759-92-7 |
|---|---|
| Molecular Formula | Cl3Eu·6H2O |
| Molecular Weight (g/mol) | 366.4 |
| MDL Number | MFCD00149701 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC199600050
|
Thermo Scientific Chemicals
199600050 |
5 g | Glass bottle |
Each for $264.30
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
| 13759-92-7 | |
| Cl3Eu·6H2O | |
| 366.4 | |
| MFCD00149701 |
Specifications
| 850.0°C | |
| 13759-92-7 | |
| 5 g | |
| Cl3Eu·6H2O | |
| MFCD00149701 | |
| Solubility in water: soluble | |
| 366.4 | |
| Trace Metal Basis | |
| 4.89 | |
| Europium(III) chloride hexahydrate, 99.9% |
| White | |
| Crystals | |
| 99.90% | |
| EuCl3·6H2O | |
| 10, 3854 | |
| 366.4 | |
| 99.9% | |
| Glass bottle | |
| 4.8900g/mL |
Safety and Handling
HYGROSCOPIC
RUO – Research Use Only