missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) bromide hydrate, REacton™, 99.99% (REO)

EuBr3·xH2O

$245.38 - $245.38

Chemical Identifiers

Molecular Formula EuBr3·xH2O
MDL Number MFCD00049921
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1321306
View Documents
Thermo Scientific Chemicals
01321306
5 g
Each for $245.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

EuBr3·xH2O
MFCD00049921

Specifications

White
5 g
EuBr3·xH2O
Soluble in water.
Odorless
Europium(III) bromide hydrate
Crystals
99.99% (REO)
MFCD00049921
391.69 (Anhydrous)
Hygroscopic
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 237-349-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only