missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) bromide hydrate, REacton™, 99.99% (REO)

Catalog No. AA1321306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1321306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1321306 Supplier Thermo Scientific Chemicals Supplier No. 01321306
Only null left

EuBr3·xH2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula EuBr3·xH2O
MDL Number MFCD00049921

Specifications

Color White
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 391.69 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Europium(III) bromide hydrate
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 237-349-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only