Learn More
Globe Scientific Euromex USB-3 Cameras
Euromex high-speed USB-3 cameras with ImageFocus Alpha capture and analysis software are the perfect solutions for todayâs educational, laboratory and industrial microscopy needs. The cameras are suitable for life science, material science and stereo microscopes.
Supplier: Globe Scientific EDC5000PRO
Description
- The CMEX-5 Pro, CMEX-10 Pro and CMEX-18 Pro cameras are equipped with relatively 5.1, 10 or 18MP CMOS sensors with 12-bits grayscale conversion and a 24-bits color rendering. li>These cameras are equipped with a USB-3 data interface enabling fast frame rates.
- Besides taking images and videos, the ImageFocus Alpha software allows measurements on still and live images and to perform annotations on captured images.
- Compatible with Windows 10 and 11, both 32- and 64-bits configurations.
- A 2GB RAM 2.8GHz computer and 17-inch display is recommended as a minimum requirement.
- High speed cameras
- ADC 12-bits, color depth 24-bits
- Excellent software
- USB 3.0 interface (USB 2.0 compatible)
- 1 to 18MP CMOS sensors
- Great color reproduction
- Low signal/noise ratio
Specifications
| Euormex CMEX-5 Pro, 5.0MP Digital USB-3 Camera | |
| 5.0MP Digital | |
| 110/220 V |
| Not Autoclavable | |
| Non-sterile |