Learn More
Eisco™ Euro-Design Balance Weights
For classroom use.
Supplier:  Eisco™ PHBWUR2
 View more versions of this product
Catalog No. S24435
Only null left
Description
Description
  • Made from stainless-steel
  • Supplied in wooden block
  • With marked weight
Specifications
Chemical Identifiers
Balance Weight Set, Euro Design
Stainless Steel
Wood
Specifications
Balance Weight Set, Euro Design
Wood
1 x 500 g, 1 x 200 g, 2 x 100 g, 1 x 50 g, 1 x 20 g, 2 x 10 g, 1 x 5 g, 2 x 2 g, 1 x 1 g
Stainless Steel
1000 g
Product Suggestions
Product Suggestions