missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Eugenol

Catalog No. E51791100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E51791100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E51791100G Supplier Sigma Aldrich Supplier No. E51791100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Eugenol

Specifications

CAS 97-53-0
Melting Point -12°C to +10°C (lit.)
Density 1.067 g/mL (at 25°C (literature))
Boiling Point 254°C (lit.)
Molecular Formula C10H12O2
Linear Formula 4-(H2C=CHCH2)C6H3-2-(OCH3)OH
Refractive Index n20/D 1.541 (literature)
MDL Number MFCD00008654
Quantity 100 g
Synonym 2-Methoxy-4-(2-propenyl)phenol; 4-Allyl-2-methoxyphenol; 4-Allylguaiacol
Molecular Weight (g/mol) 164.2
Percent Purity 99%
Show More Show Less