missing translation for 'onlineSavingsMsg'
Learn More

Etoposide 98.0+%, TCI America™

Catalog No. E0675100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0675100MG 100 mg
E06751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0675100MG Supplier TCI America Supplier No. E0675100MG

Chemical Identifiers

CAS 33419-42-0
Molecular Formula C29H32O13
Molecular Weight (g/mol) 588.56
MDL Number MFCD00869325,MFCD00869325
InChI Key VJJPUSNTGOMMGY-MRVIYFEKSA-N
Synonym vjjpusntgommgy-nzlmilqcsa
PubChem CID 50936917
IUPAC Name (10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
SMILES COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12

Specifications

Melting Point 251°C
Color White
Quantity 100 mg
Formula Weight 588.56
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Etoposide
RTECSNumber KC0190000
TSCA No