missing translation for 'onlineSavingsMsg'
Learn More

Etodolac 98.0+%, TCI America™

Supplier:  TCI America E085825G

 View more versions of this product

Catalog No. E085825G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Etodolac
148°C
C17H21NO3
25 g
etodolac, etodolic acid, lodine, ultradol, lodine xl, etodolaco, etodolacum, ramodar, etodolacum inn-latin, etodolaco inn-spanish
CCC1=C2NC3=C(CCOC3(CC)CC(O)=O)C2=CC=C1
287.36
CHEBI:4909
≥98.0% (HPLC,T)
41340-25-4
Yellow
MFCD00133313
2811
NNYBQONXHNTVIJ-UHFFFAOYNA-N
2-{1,8-diethyl-1H,3H,4H,9H-pyrano[3,4-b]indol-1-yl}acetic acid
3308
287.36
Crystalline Powder

Chemical Identifiers

41340-25-4
287.36
NNYBQONXHNTVIJ-UHFFFAOYNA-N
3308
2-{1,8-diethyl-1H,3H,4H,9H-pyrano[3,4-b]indol-1-yl}acetic acid
C17H21NO3
MFCD00133313
etodolac, etodolic acid, lodine, ultradol, lodine xl, etodolaco, etodolacum, ramodar, etodolacum inn-latin, etodolaco inn-spanish
CHEBI:4909
CCC1=C2NC3=C(CCOC3(CC)CC(O)=O)C2=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UQ0360000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions