missing translation for 'onlineSavingsMsg'
Learn More

Etodolac 98.0+%, TCI America™

Catalog No. E08585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08585G 5 g
E085825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08585G Supplier TCI America Supplier No. E08585G

Chemical Identifiers

CAS 41340-25-4
Molecular Formula C17H21NO3
Molecular Weight (g/mol) 287.36
MDL Number MFCD00133313
InChI Key NNYBQONXHNTVIJ-UHFFFAOYNA-N
Synonym etodolac, etodolic acid, lodine, ultradol, lodine xl, etodolaco, etodolacum, ramodar, etodolacum inn-latin, etodolaco inn-spanish
PubChem CID 3308
ChEBI CHEBI:4909
IUPAC Name 2-{1,8-diethyl-1H,3H,4H,9H-pyrano[3,4-b]indol-1-yl}acetic acid
SMILES CCC1=C2NC3=C(CCOC3(CC)CC(O)=O)C2=CC=C1

Specifications

Melting Point 148°C
Color Yellow
Quantity 5 g
UN Number 2811
Formula Weight 287.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Etodolac
RTECSNumber UQ0360000
TSCA No
Recommended Storage Refrigerator