missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Etodolac

Chemical Identifiers

CAS 41340-25-4
Chemical Name or Material Etodolac
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC465720050
View Documents
Thermo Scientific™
465720050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC465720250
View Documents
Thermo Scientific™
465720250
25 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

41340-25-4
Etodolac

Specifications

41340-25-4
5 g
150.0°C to 152.0°C
Etodolac
Videos
Safety and Handling

Safety and Handling

GHS H Statement Toxic if swallowed.

Causes serious eye irritation.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only