missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etidronic Acid, Approx. 60%, Spectrum™ Chemical
E3490, 2809-21-4, C2H8O7P2
$889.54 - $889.54
Chemical Identifiers
| CAS | 2809-21-4 |
|---|---|
| Molecular Formula | C2H8O7P2 |
| Molecular Weight (g/mol) | 206.03 |
| InChI Key | DBVJJBKOTRCVKF-UHFFFAOYSA-N |
| IUPAC Name | (1-hydroxy-1-phosphonoethyl)phosphonic acid |
| SMILES | CC(O)(P(O)(O)=O)P(O)(O)=O |
Description
Spectrum™ Chemical Etidronic Acid , also known as 1-hydroxyethane 1,1-diphosphonic acid, is used as a bisphosphonate to treat osteoporosis. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 2809-21-4 | |
| 206.03 | |
| (1-hydroxy-1-phosphonoethyl)phosphonic acid |
| C2H8O7P2 | |
| DBVJJBKOTRCVKF-UHFFFAOYSA-N | |
| CC(O)(P(O)(O)=O)P(O)(O)=O |
Specifications
| 2809-21-4 | |
| Amber Glass Bottle | |
| 500 g | |
| CC(O)(P(O)(O)=O)P(O)(O)=O | |
| 206.03 | |
| Reagent |
| 100% | |
| C2H8O7P2 | |
| DBVJJBKOTRCVKF-UHFFFAOYSA-N | |
| (1-hydroxy-1-phosphonoethyl)phosphonic acid | |
| ∼60% |