missing translation for 'onlineSavingsMsg'
Learn More

Ethynylestradiol 98.0+%, TCI America™

Catalog No. E00371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E00371G Supplier TCI America Supplier No. E00371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-63-6
Molecular Formula C20H24O2
Molecular Weight (g/mol) 296.41
MDL Number MFCD00003690
InChI Key BFPYWIDHMRZLRN-SLHNCBLASA-N
Synonym ethinyl estradiol, ethynylestradiol, ethynyl estradiol, ethinylestradiol, ethinyloestradiol, ginestrene, 17-ethinylestradiol, progynon c, ethinoral, eticyclin
PubChem CID 5991
ChEBI CHEBI:4903
IUPAC Name (1R,3aS,3bR,9bS,11aS)-1-ethynyl-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Melting Point 182°C
Quantity 1 g
Formula Weight 296.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethynylestradiol
RTECSNumber RC8925000
TSCA Yes