missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriethoxysilane, 96%

Catalog No. AAL16495AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL16495AC 25 mL
AAL16495AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL16495AC Supplier Thermo Scientific Chemicals Supplier No. L16495AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 78-07-9 | C8H20O3Si | 192.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-07-9
Molecular Formula C8H20O3Si
Molecular Weight (g/mol) 192.33
MDL Number MFCD00026754
InChI Key DENFJSAFJTVPJR-UHFFFAOYSA-N
Synonym ethyltriethoxysilane, triethoxy ethyl silane, silane, triethoxyethyl, a 15 silane, union carbide a-15, triethoxy-ethylsilane, silane, ethyltriethoxy, silane, ethyl triethoxy a-15, ethaneorthosiliconic acid, triethyl ester, ethyl triethoxysilane
PubChem CID 6515
IUPAC Name triethoxy(ethyl)silane
SMILES CCO[Si](CC)(OCC)OCC

Specifications

Boiling Point 158°C to 159°C
Melting Point -78°C
Quantity 25 mL
UN Number UN1993
Beilstein 1362028
Flash Point 40°C (104°F)
Formula Weight 192.33
Percent Purity 96%
Refractive Index 1.392
Sensitivity Moisture sensitive
Density 0.896
Chemical Name or Material Ethyltriethoxysilane
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 201-080-1
RTECSNumber VV4205000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only