missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyltrichlorosilane 98.0+%, TCI America™
$53.90 - $53.90
Chemical Identifiers
| CAS | 115-21-9 |
|---|---|
| Molecular Formula | C2H5Cl3Si |
| Molecular Weight (g/mol) | 163.497 |
| MDL Number | MFCD00000485 |
| InChI Key | ZOYFEXPFPVDYIS-UHFFFAOYSA-N |
| Synonym | ethyltrichlorosilane, trichloro ethyl silane, trichloroethylsilicane, silane, trichloroethyl, ethyl silicon trichloride, trichloroethylsilicon, silicane, trichloroethyl, ethyl trichlorosilane, silane, ethyltrichloro, c2h5sicl3 |
| PubChem CID | 8260 |
| IUPAC Name | trichloro(ethyl)silane |
| SMILES | CC[Si](Cl)(Cl)Cl |
Chemical Identifiers
| 115-21-9 | |
| 163.497 | |
| ZOYFEXPFPVDYIS-UHFFFAOYSA-N | |
| 8260 | |
| CC[Si](Cl)(Cl)Cl |
| C2H5Cl3Si | |
| MFCD00000485 | |
| ethyltrichlorosilane, trichloro ethyl silane, trichloroethylsilicane, silane, trichloroethyl, ethyl silicon trichloride, trichloroethylsilicon, silicane, trichloroethyl, ethyl trichlorosilane, silane, ethyltrichloro, c2h5sicl3 | |
| trichloro(ethyl)silane |
Specifications
| 115-21-9 | |
| 100°C | |
| MFCD00000485 | |
| 1196 | |
| ZOYFEXPFPVDYIS-UHFFFAOYSA-N | |
| trichloro(ethyl)silane | |
| 8260 | |
| ≥98.0% (GC,T) | |
| Ethyltrichlorosilane |
| Yellow | |
| C2H5Cl3Si | |
| 25 g | |
| ethyltrichlorosilane, trichloro ethyl silane, trichloroethylsilicane, silane, trichloroethyl, ethyl silicon trichloride, trichloroethylsilicon, silicane, trichloroethyl, ethyl trichlorosilane, silane, ethyltrichloro, c2h5sicl3 | |
| CC[Si](Cl)(Cl)Cl | |
| 163.497 | |
| 163.50 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2041
RTECSNumber : VV4200000
TSCA : Yes