missing translation for 'onlineSavingsMsg'
Learn More

Ethylmalonic acid, 97+%

CAS: 601-75-2 | C5H8O4 | 132.115 g/mol

$80.79 - $847.85

Chemical Identifiers

CAS 601-75-2
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00002668
InChI Key UKFXDFUAPNAMPJ-UHFFFAOYSA-N
Synonym ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid
PubChem CID 11756
ChEBI CHEBI:741548
IUPAC Name 2-ethylpropanedioic acid
SMILES CCC(C(=O)O)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2071506
View Documents
Thermo Scientific Chemicals
B2071506
5 g
Each for $80.79
Only null left
 
AAB2071514
View Documents
Thermo Scientific Chemicals
B2071514
25 g
Each for $269.85
Only null left
 
AAB2071522
View Documents
Thermo Scientific Chemicals
B2071522
100 g
Each for $847.85
Only null left
 
Description

Description

Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water, methanol (very faint turbidity), and ethanol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

601-75-2
132.115
UKFXDFUAPNAMPJ-UHFFFAOYSA-N
11756
2-ethylpropanedioic acid
C5H8O4
MFCD00002668
ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid
CHEBI:741548
CCC(C(=O)O)C(=O)O

Specifications

601-75-2
C5H8O4
5 g
ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid
UKFXDFUAPNAMPJ-UHFFFAOYSA-N
2-ethylpropanedioic acid
11756
132.12
Ethylmalonic acid
112°C to 114°C
MFCD00002668
774334
Soluble in water,methanol (very faint turbidity),and ethanol.
CCC(C(=O)O)C(=O)O
132.115
CHEBI:741548
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 210-007-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only