missing translation for 'onlineSavingsMsg'
Learn More

Ethylmalonic acid, 97+%

Catalog No. AAB2071506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2071506 5 g
AAB2071514 25 g
AAB2071522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2071506 Supplier Thermo Scientific Chemicals Supplier No. B2071506

CAS: 601-75-2 | C5H8O4 | 132.115 g/mol

Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water, methanol (very faint turbidity), and ethanol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 601-75-2
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00002668
InChI Key UKFXDFUAPNAMPJ-UHFFFAOYSA-N
Synonym ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid
PubChem CID 11756
ChEBI CHEBI:741548
IUPAC Name 2-ethylpropanedioic acid
SMILES CCC(C(=O)O)C(=O)O

Specifications

Melting Point 112°C to 114°C
Quantity 5 g
Beilstein 774334
Solubility Information Soluble in water,methanol (very faint turbidity),and ethanol.
Formula Weight 132.12
Percent Purity 98%
Chemical Name or Material Ethylmalonic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-007-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only