missing translation for 'onlineSavingsMsg'
Learn More

Ethylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. p-7051629
Change view
Click to view available options
Quantity:
0.5 mol
1 mol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41675JT 0.5 mol
AA41675JU 1 mol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41675JT Supplier Thermo Scientific Chemicals Supplier No. 041675JT
Only null left

CAS: 925-90-6 | C2H5BrMg | 133.27 g/mol

Ethylmagnesium bromide is used as a reagent and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 925-90-6
Molecular Formula C2H5BrMg
Molecular Weight (g/mol) 133.27
MDL Number MFCD00000043
InChI Key TWTWFMUQSOFTRN-UHFFFAOYSA-M
Synonym ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent
PubChem CID 101914
IUPAC Name magnesium;ethane;bromide
SMILES CC[Mg]Br

Specifications

Melting Point -116.3°C
Color Brown to Yellow
Density 1.020 g/mL
Boiling Point 34.6°C
Odor Ether-like
Packaging ChemSeal™ bottles
Linear Formula C2H5MgBr
Quantity 0.5 mol
UN Number UN3399
Sensitivity Air and moisture sensitive; May form a precipitate
Solubility Information Miscible in alcohols,ketones,esters,ethers,hydrocarbons.
Formula Weight 133.28
Concentration or Composition (by Analyte or Components) 3M in ether
Physical Form Liquid
Chemical Name or Material Ethylmagnesium bromide
Show More Show Less
Hazard Category H224-H260-H302-H314-H336
Hazard Statement GHS H Statement
H224-H260-H314-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
EINECSNumber 213-127-3
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only