missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylhexadecyldimethylammonium Bromide 98.0+%, TCI America™
$59.65 - $436.19
Chemical Identifiers
| CAS | 124-03-8 |
|---|---|
| Molecular Formula | C20H44BrN |
| Molecular Weight (g/mol) | 378.483 |
| MDL Number | MFCD00011774 |
| InChI Key | VUFOSBDICLTFMS-UHFFFAOYSA-M |
| Synonym | cetyldimethylethylammonium bromide, ethylhexadecyldimethylammonium bromide, cetylcide, bretol, ethyl cetab, ammonyx dme, cetethyldimonium bromide, n-ethyl-n,n-dimethylhexadecan-1-aminium bromide, dimethylethylhexadecylammonium bromide, quaternium 17 |
| PubChem CID | 31280 |
| IUPAC Name | ethyl-hexadecyl-dimethylazanium;bromide |
| SMILES | CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-] |
Chemical Identifiers
| 124-03-8 | |
| 378.483 | |
| VUFOSBDICLTFMS-UHFFFAOYSA-M | |
| 31280 | |
| CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-] |
| C20H44BrN | |
| MFCD00011774 | |
| cetyldimethylethylammonium bromide, ethylhexadecyldimethylammonium bromide, cetylcide, bretol, ethyl cetab, ammonyx dme, cetethyldimonium bromide, n-ethyl-n,n-dimethylhexadecan-1-aminium bromide, dimethylethylhexadecylammonium bromide, quaternium 17 | |
| ethyl-hexadecyl-dimethylazanium;bromide |
Specifications
| 124-03-8 | |
| C20H44BrN | |
| 25 g | |
| VUFOSBDICLTFMS-UHFFFAOYSA-M | |
| ethyl-hexadecyl-dimethylazanium;bromide | |
| 31280 | |
| ≥98.0% (T) | |
| Ethylhexadecyldimethylammonium Bromide |
| White | |
| MFCD00011774 | |
| cetyldimethylethylammonium bromide, ethylhexadecyldimethylammonium bromide, cetylcide, bretol, ethyl cetab, ammonyx dme, cetethyldimonium bromide, n-ethyl-n,n-dimethylhexadecan-1-aminium bromide, dimethylethylhexadecylammonium bromide, quaternium 17 | |
| CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-] | |
| 378.483 | |
| 378.48 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0184&(9)-1971
RTECSNumber : BQ5425000
TSCA : Yes