missing translation for 'onlineSavingsMsg'
Learn More

Ethylhexadecyldimethylammonium Bromide 98.0+%, TCI America™

Catalog No. D072125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D072125G 25 g
D0721500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D072125G Supplier TCI America Supplier No. D072125G

Chemical Identifiers

CAS 124-03-8
Molecular Formula C20H44BrN
Molecular Weight (g/mol) 378.483
MDL Number MFCD00011774
InChI Key VUFOSBDICLTFMS-UHFFFAOYSA-M
Synonym cetyldimethylethylammonium bromide, ethylhexadecyldimethylammonium bromide, cetylcide, bretol, ethyl cetab, ammonyx dme, cetethyldimonium bromide, n-ethyl-n,n-dimethylhexadecan-1-aminium bromide, dimethylethylhexadecylammonium bromide, quaternium 17
PubChem CID 31280
IUPAC Name ethyl-hexadecyl-dimethylazanium;bromide
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-]

Specifications

Color White
Quantity 25 g
Formula Weight 378.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylhexadecyldimethylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
RTECSNumber BQ5425000
TSCA Yes