missing translation for 'onlineSavingsMsg'
Learn More

(Ethylenedinitrilo)tetraacetic Acid Disodium, Dihydrate, Reagent, ACS, 99-101%, Spectrum™ Chemical

Catalog No. p-7157373
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600004 125 g Poly Bottle
18600006 500 g Poly Bottle
18600005 6 x 125 g Poly Bottle
18600008 2.5 kg Poly Bottle
18600007 6 x 500 g Poly Bottle
18600009 12 kg Poly Pail
18600010 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18600004 Supplier Spectrum Chemical Mfg Cor Supplier No. E1045125GM
Only null left

E1045, 6381-92-6, C10H14N2Na2O8·2H2O

Spectrum™ Chemical (Ethylenedinitrilo)tetraacetic Acid Disodium, Dihydrate, Reagent, ACS , is one of several salts produced from Ethylenediaminetetraacetic acid (EDTA), itself an aminopolycarboxylic acid. Widely used in laboratory conditions, its uses vary from scavenging metal ions to acting as a chelating agent that binds cells in tissue cultures. As an ACS grade Reagent, Spectrum Chemical manufactured (ETHYLENEDINITRILO)TETRAACETIC ACID DISODIUM, DIHYDRATE is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 125 g
Percent Purity 99 to 101%
Grade ACS