missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Ethylenedinitrilo)tetraacetic Acid Disodium, Dihydrate, Reagent, ACS, 99-101%, Spectrum™ Chemical
Description
Spectrum™ Chemical (Ethylenedinitrilo)tetraacetic Acid Disodium, Dihydrate, Reagent, ACS , is one of several salts produced from Ethylenediaminetetraacetic acid (EDTA), itself an aminopolycarboxylic acid. Widely used in laboratory conditions, its uses vary from scavenging metal ions to acting as a chelating agent that binds cells in tissue cultures. As an ACS grade Reagent, Spectrum Chemical manufactured (ETHYLENEDINITRILO)TETRAACETIC ACID DISODIUM, DIHYDRATE is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Pail |
| Quantity | 12 kg |
| Percent Purity | 99 to 101% |
| Grade | ACS |