missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Trisodium Salt Monohydrate, 99.32%, MP Biomedicals™

Catalog No. ICN15108880
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15108880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15108880 Supplier MP Biomedicals Inc Supplier No. 0215108880
Only null left

Chemical Identifiers

CAS 10378-22-0
Molecular Formula C10H18N2Na3O9
Molecular Weight (g/mol) 379.228
MDL Number MFCD00149675
InChI Key DFCJTQJARZUURB-UHFFFAOYSA-N
Synonym edta, trisodium salt trihydrate
PubChem CID 131865091
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;hydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.[Na].[Na].[Na]

Specifications

Color White
Quantity 100 g
Formula Weight 376.2
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate