missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate, MP Biomedicals™

Catalog No. p-4774296
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15108890 500 g
ICN15108891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15108890 Supplier MP Biomedicals Inc Supplier No. 0215108890
Only null left

Water soluble. Clear colorless to light yellow solution at 32gm plus 100mL of 1M sodium hydroxide

Chemical Identifiers

CAS 85715-60-2
Molecular Formula C10H18N2Na3O9
Molecular Weight (g/mol) 379.228
MDL Number MFCD00149675
InChI Key DFCJTQJARZUURB-UHFFFAOYSA-N
Synonym edta, trisodium salt trihydrate
PubChem CID 131865091
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;hydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.[Na].[Na].[Na]

Specifications

Color White
Quantity 500 g
Formula Weight 358.19
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate
EINECSNumber 200-449-4