missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%
CAS: 65501-24-8 | C10H13K3N2O8·2H2O | 442.56 g/mol
$230.23 - $230.23
Chemical Identifiers
| CAS | 65501-24-8 |
|---|---|
| Molecular Formula | C10H13K3N2O8·2H2O |
| Molecular Weight (g/mol) | 442.56 |
| InChI Key | MAPFUJCWRWFQIY-UHFFFAOYSA-K |
| Synonym | ethylenediaminetetraacetic acid tripotassium salt dihydrate, edta tripotassium salt, edta tripotassium salt dihydrate, tripotassium ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-carboxylatomethyl carboxymethyl amino ethyl azanediyl diacetate dihydrate, tripotassium 2-2-bis carboxylatomethyl amino ethyl carboxymethyl amino acetate dihydrate, edta tripotassium salt, dihydrate, ethylenediaminetetraacetic acid, tripotassium salt dihydrate, ethylenediaminetetraacetic acid tripotassium salt dihydrate titration |
| PubChem CID | 2733162 |
| IUPAC Name | tripotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;dihydrate |
| SMILES | C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].O.O.[K+].[K+].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC301601000
|
Thermo Scientific Chemicals
301601000 |
100 g | Plastic bottle |
Each for $230.23
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentChemical Identifiers
| 65501-24-8 | |
| 442.56 | |
| ethylenediaminetetraacetic acid tripotassium salt dihydrate, edta tripotassium salt, edta tripotassium salt dihydrate, tripotassium ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-carboxylatomethyl carboxymethyl amino ethyl azanediyl diacetate dihydrate, tripotassium 2-2-bis carboxylatomethyl amino ethyl carboxymethyl amino acetate dihydrate, edta tripotassium salt, dihydrate, ethylenediaminetetraacetic acid, tripotassium salt dihydrate, ethylenediaminetetraacetic acid tripotassium salt dihydrate titration | |
| tripotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;dihydrate |
| C10H13K3N2O8·2H2O | |
| MAPFUJCWRWFQIY-UHFFFAOYSA-K | |
| 2733162 | |
| C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].O.O.[K+].[K+].[K+] |
Specifications
| 65501-24-8 | |
| Authentic | |
| Plastic bottle | |
| (KOCOCH2)2NCH2CH2N(CH2COOK)CH2COOH·2H2O | |
| ethylenediaminetetraacetic acid tripotassium salt dihydrate, edta tripotassium salt, edta tripotassium salt dihydrate, tripotassium ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-carboxylatomethyl carboxymethyl amino ethyl azanediyl diacetate dihydrate, tripotassium 2-2-bis carboxylatomethyl amino ethyl carboxymethyl amino acetate dihydrate, edta tripotassium salt, dihydrate, ethylenediaminetetraacetic acid, tripotassium salt dihydrate, ethylenediaminetetraacetic acid tripotassium salt dihydrate titration | |
| C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].O.O.[K+].[K+].[K+] | |
| 442.56 | |
| 442.56 | |
| Fine Crystalline Powder |
| White | |
| 99% min. (on dry substance) (Complexometry) | |
| C10H13K3N2O8·2H2O | |
| 100 g | |
| MAPFUJCWRWFQIY-UHFFFAOYSA-K | |
| tripotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;dihydrate | |
| 2733162 | |
| 99+% | |
| Ethylenediaminetetraacetic acid, tripotassium salt dihydrate |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only