Learn More
Ethylenediaminetetraacetic acid tetrasodium salt hydrate, 98%
CAS: 194491-31-1 | C10H12N2Na4O8 | 380.17 g/mol
$54.73 - $455.45
Chemical Identifiers
| CAS | 194491-31-1 |
|---|---|
| Molecular Formula | C10H12N2Na4O8 |
| Molecular Weight (g/mol) | 380.17 |
| MDL Number | MFCD00150025 |
| InChI Key | UEUXEKPTXMALOB-UHFFFAOYSA-J |
| Synonym | edta tetrasodium salt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, tetrasodium 2-2-bis carboxylatomethyl amino ethyl carboxylatomethyl amino acetate hydrate, tetrasodium ion 4-edta hydrate, ethylenediaminetetraacetic acid, tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, bioultra kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, practical grade |
| PubChem CID | 16211056 |
| IUPAC Name | tetrasodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;hydrate |
| SMILES | [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1738530
|
Thermo Scientific Chemicals
A1738530 |
250 g |
Each for $54.73
|
|
|||||
|
AAA173850B
|
Thermo Scientific Chemicals
A173850B |
1000 g |
Each for $133.81
|
|
|||||
|
AAA173850I
|
Thermo Scientific Chemicals
A173850I |
5000 g |
Each for $455.45
|
|
|||||
Description
Ethylenediaminetetraacetic acid tetrasodium salt hydrate is used as an inhibitor of metal ligand proteases. It also acts as a chelator of zinc ion in the active site of metalloproteases. It is used as an anticoagulant and also used in the fluorometric determination of beryllium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylenediaminetetraacetic acid tetrasodium salt hydrate is used as an inhibitor of metal ligand proteases. It also acts as a chelator of zinc ion in the active site of metalloproteases. It is used as an anticoagulant and also used in the fluorometric determination of beryllium.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and metals.
Chemical Identifiers
| 194491-31-1 | |
| 380.17 | |
| UEUXEKPTXMALOB-UHFFFAOYSA-J | |
| 16211056 | |
| [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O |
| C10H12N2Na4O8 | |
| MFCD00150025 | |
| edta tetrasodium salt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, tetrasodium 2-2-bis carboxylatomethyl amino ethyl carboxylatomethyl amino acetate hydrate, tetrasodium ion 4-edta hydrate, ethylenediaminetetraacetic acid, tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, bioultra kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, practical grade | |
| tetrasodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;hydrate |
Specifications
| 194491-31-1 | |
| C10H12N2Na4O8 | |
| 250 g | |
| edta tetrasodium salt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, tetrasodium 2-2-bis carboxylatomethyl amino ethyl carboxylatomethyl amino acetate hydrate, tetrasodium ion 4-edta hydrate, ethylenediaminetetraacetic acid, tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, bioultra kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, practical grade | |
| UEUXEKPTXMALOB-UHFFFAOYSA-J | |
| tetrasodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;hydrate | |
| 16211056 | |
| 98% |
| >300°C | |
| MFCD00150025 | |
| 3828865 | |
| Soluble in water. | |
| [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O | |
| 380.17 | |
| 380.17 (Anhydrous) | |
| Ethylenediaminetetraacetic acid tetrasodium salt hydrate |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
H302+H332-H318-H335
EINECSNumber : 200-573-9
RTECSNumber : AH5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only