missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tetrasodium salt hydrate, 98%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
Specifications
Specifications
| Melting Point | >300.0°C |
| Color | White to Yellow |
| pH | 9.5 to 10.5 (1% soln.) |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Assay Percent Range | 97.5% min. (on dry substance) (Complexometry) |
| Linear Formula | (NaO2CCH2)2NCH2CH2N(CH2CO2Na)2·xH2O |
| Fieser | 01,373; 05,296 |
| Merck Index | 14,3517 |
| Solubility Information | Solubility in water: 800g/L (20°C) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye damage. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
RUO – Research Use Only