missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Magnesium Disodium Salt, Spectrum™ Chemical

Catalog No. 18610450
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610450 100 g Poly Bottle
18610451 250 g Poly Bottle
18610452 1 kg Poly Pail
18610453 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610450 Supplier Spectrum Chemical Mfg Cor Supplier No. E1044100GM

Chemical Identifiers

CAS 14402-88-1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Grade Reagent
Insoluble Matter 5e-05

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.