missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Magnesium Disodium Salt Hydrate 99.0+%, TCI America™

Catalog No. E009425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E009425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E009425G Supplier TCI America Supplier No. E009425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity magnesium disodium EDTA derivative for chelating and complexation applications.

Chemical Identifiers

CAS 14402-88-1
Molecular Formula C10H12MgN2Na2O8
Molecular Weight (g/mol) 358.50
MDL Number MFCD00078217
InChI Key AWNVVAMWLMUZOZ-UHFFFAOYSA-J
Synonym unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate
PubChem CID 161064
IUPAC Name magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
SMILES [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 358.50
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Magnesium Disodium Salt Hydrate
EINECSNumber (2)-1265
TSCA Yes