missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure

Catalog No. AC304680010
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC304680010 1 kg
AC304682500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC304680010 Supplier Thermo Scientific Chemicals Supplier No. 304680010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, C10H12FeN2NaO8.3H2O, CAS Number-18154-32-, sodium ironedetate, edta ferric sodium salt, calmosine, sodium feredetate, edta iron iii sodium salt, sequestrene nafe, ferric sodium edta, ferisan, sodium iron edta, sytron, 1kg | CAS: 18154-32-0 | C10H12FeN2NaO8·3H2O | 421.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 18154-32-0
Molecular Formula C10H12FeN2NaO8·3H2O
Molecular Weight (g/mol) 421.1
MDL Number MFCD00798117
InChI Key MKWYFZFMAMBPQK-UHFFFAOYSA-J
Synonym sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edta
PubChem CID 27461
ChEBI CHEBI:78292
IUPAC Name sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+)
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3]

Specifications

Color Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 1 kg
Merck Index 15,4058
Solubility Information Solubility in water: soluble.
Formula Weight 421.1
Percent Purity 98%
Grade Pure
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only