missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure

Catalog No. p-7082789
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Nalgene bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ15694AE 100 mL Nalgene bottle
AAJ15694AP 500 mL Nalgene bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ15694AE Supplier Thermo Fisher Scientific Supplier No. J15694AE
Only null left

Chelating agent used in many enzyme buffers and at higher concentrations, as enzyme inactivator | CAS: 6381-92-6 | C10H18N2Na2O10 | 372.238 g/mol

Chelating agent used in many enzyme buffers and, at higher concentrations, as an enzyme inactivator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.238
MDL Number MFCD00150037
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]

Specifications

Packaging Nalgene bottle
Quantity 100 mL
Beilstein 3900609
Grade Molecular Biology, Ultrapure
Concentration or Composition (by Analyte or Components) 0.5 M
Physical Form Solution
Chemical Name or Material Ethylenediaminetetraacetic acid
Hazard Category H373
Precautionary Statement P260-P314-P501c
EINECSNumber 205-358-3
RTECSNumber AH4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only