missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Disodium Zinc Salt Hydrate 98.0+%, TCI America™

Catalog No. p-7128836
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E009825G 25 g
E0098500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E009825G Supplier TCI America Supplier No. E009825G
Only null left

Chemical Identifiers

CAS 14025-21-9
Molecular Formula C10H12N2Na2O8Zn
Molecular Weight (g/mol) 399.57
MDL Number MFCD00135187
InChI Key UBLOJEHIINPTTG-UHFFFAOYSA-J
Synonym Disodium Zinc Ethylenediaminetetraacetate
PubChem CID 51808
IUPAC Name zinc(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
SMILES [Na+].[Na+].[Zn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 399.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Disodium Zinc Salt Hydrate
EINECSNumber (2)-1266
TSCA Yes