Learn More
Honeywell Fluka™ EDTA disodium salt solution with zinc complex added (solution A), Volumetric, for water hardness determination, Honeywell Fluka™
Volumetric, for water hardness determination (1 ml = 5.6 German degree of hardness in 100 ml of water)
$410.87
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
6002531
View Documents Promotion Details
Honeywell Fluka™
345471L
1 L
Each for $410.87
Only null left
Description
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations