missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate, Honeywell Fluka™

Catalog No. 6001818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001818 10 g
6001819 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001818 Supplier Honeywell Chemicals Supplier No. 0368210G
Only null left

for HPLC,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Quantity 10 g
UN Number NONH for all modes of transport
Beilstein 3900609
Flash Point >100°C (>212°F)
Solubility Information Soluble in water (0.1M)
Formula Weight 372.24g/mol
Grade HPLC
Chemical Name or Material Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate
Hazard Statement P260
Precautionary Statement H332-H373
EINECSNumber 205-358-3
RTECSNumber AH4410000