Learn More
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical S31110
Specifications
| Ethylenediamine Tetraacetic Acid, Disodium Salt Dihydrate | |
| >252°C | |
| White | |
| 99.0 to 101.0% | |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
| 44120005 | |
| 99.0 to 101.0% | |
| Solid |
| 6381-92-6 | |
| 252°C | |
| 4 to 6 | |
| Poly Pail | |
| (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O | |
| 10 kg | |
| 0.005% max. Insoluble Matter | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 372.24 | |
| 372.24 | |
| Certified ACS |
Chemical Identifiers
| 6381-92-6 | |
| 372.24 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| 44120005 | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:0