missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
Specifications
Specifications
| Color | White |
| Assay Percent Range | ≥99% |
| Quantity | 500 g |
| IUPAC Name | disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
| Formula Weight | 372.2 |
| Percent Purity | ≥99% |
| Grade | Molecular Biology Reagent |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate |
Safety and Handling
| EINECSNumber | 205-358-3 |
| Recommended Storage | Store at Room Temperature (15 to 30°C). |