missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. AC327201000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327201000 100 g Plastic bottle
AC327205000 500 g Plastic bottle
AC327200025 2.5 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC327201000 Supplier Thermo Scientific Chemicals Supplier No. 327201000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6381-92-6 | C10H18N2Na2O10 | 372.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent for molecular biology

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Absorbance (0.1M in water), (0.1 M in water) at 0nm, 0.02 max. at 280nm, 0.2 max. at 260nm
Melting Point 252.0°C
Color White
pH 4 to 5 (5% soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Complexometry)
Packaging Plastic bottle
Quantity 100 g
Merck Index 153565
Solubility Information Solubility in water: freely soluble. Other solubilities: slightly soluble in alcohol (95%),insoluble in chloroform and ether
Formula Weight 372.23
Percent Purity 99+%
Grade Molecular Biology
Insoluble Matter 0.005% max
Physical Form Crystalline Solid
Chemical Name or Material Ethylenediaminetetraacetic acid, disodium salt dihydrate
Show More Show Less
Hazard Category Acute toxicity (category 4), Specific target organ toxicity after repeated exposure (category 2)
Hazard Statement Harmful if inhaled, May cause damage to organs through prolonged or repeated exposure if inhaled
EINECSNumber 200-449-4
RTECSNumber AH4025000
TSCA TSCA

RUO – Research Use Only