missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis

Catalog No. AC147855000
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147855000 500 g Plastic bottle
AC147851000 100 g Plastic bottle
AC147850010 1 kg Plastic bottle
AC147850050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC147855000 Supplier Thermo Scientific Chemicals Supplier No. 147855000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biological buffer, Chelation/Complexation reagent | CAS: 6381-92-6 | C10H18N2Na2O10 | 372.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer, Chelation/Complexation reagent

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 252.0°C
Color White
pH 4.0 to 6.0 (1% soln. at 20°C)
Loss on Drying 11.4% max. (200°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99.0 to 101% (On dry substance)
Packaging Plastic bottle
Quantity 500 g
Merck Index 15, 3565
Solubility Information Solubility in water: freely soluble. Other solubilities: slightly soluble in alcohol (95%),insoluble in chloroform and ether
Formula Weight 372.23
Percent Purity 99+%
Grade Analysis
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, disodium salt dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only