missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer, Chelation/Complexation reagent
Specifications
Specifications
| Melting Point | 252.0°C |
| Color | White |
| pH | 4.0 to 6.0 (1% soln. at 20°C) |
| Loss on Drying | 11.4% max. (200°C, 3 hrs) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.0 to 101% (On dry substance) |
| Packaging | Plastic bottle |
| Quantity | 1 kg |
| Merck Index | 15, 3565 |
| Solubility Information | Solubility in water: freely soluble. Other solubilities: slightly soluble in alcohol (95%),insoluble in chloroform and ether |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if inhaled. May cause damage to organs through prolonged or repeated exposure if inhaled. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell. |
RUO – Research Use Only