missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt concentrate with zinc complex added (solution B), For 1L standard solution, for complexometry, Honeywell Fluka™

Catalog No. p-8302130
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Ea.
Catalog No. Quantity
6002661 1 Ea.
1 options

Catalog No. 6002661

Supplier: Honeywell Chemicals 380561EA

Only null left

for 1L standard solution, 1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 139-33-3 , 14025-21-9 , 64-02-8
Molecular Formula C10H14N2Na2O8
Molecular Weight (g/mol) 336.21
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
PubChem CID 57339238
ChEBI CHEBI:64734
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 15.84, 1.19, 6.41
Quantity 1 Ea.
UN Number NONH for all modes of transport
Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt concentrate with zinc complex added (solution B)