missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Dipotassium Salt Dihydrate, 99%

Catalog No. p-7045747
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1184222 100 g
AAL1184236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1184222 Supplier Thermo Scientific Chemicals Supplier No. L1184222
Only null left

CAS: 25102-12-9 | C10H18K2N2O10 | 404.455 g/mol

Ethylenediaminetetraacetic acid dipotassium salt dihydrate is used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals. It is involved in the study of chelators, biochemicals and reagents as well as in the study of the three dimensional hierarchy structures from self-assembly of quantum dots. It also acts as a catalyst by partially removing thiol stabilizers from the nanoparticle surface to induce the stabilization reaction of CdTe nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid dipotassium salt dihydrate is used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals. It is involved in the study of chelators, biochemicals and reagents as well as in the study of the three dimensional hierarchy structures from self-assembly of quantum dots. It also acts as a catalyst by partially removing thiol stabilizers from the nanoparticle surface to induce the stabilization reaction of CdTe nanoparticles.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 25102-12-9
Molecular Formula C10H18K2N2O10
Molecular Weight (g/mol) 404.455
MDL Number MFCD00150036
InChI Key JFROQFOCFOKDKU-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid dipotassium salt dihydrate, dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-bis carboxymethyl amino ethyl azanediyl diacetate dihydrate, edta dipotassium salt dihydrate, edta dipotassium salt, ethylenediaminetetraacetic acid,dipotassium salt dihydrate, potassium ethylenediaminetetraacetate dibasic, dipotassium ethylenediaminetetraacetate dihydrate, ethylenedinitrilo tetraacetic acid dipotassium salt, acmc-209ggx
PubChem CID 53486451
IUPAC Name dipotassium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[K+].[K+]

Specifications

Melting Point ∽272°C (decomposition)
Odor Odorless
Quantity 100 g
Merck Index 14,3517
Solubility Information Soluble in water.
Formula Weight 404.46 (368.43 Anhydrous)
Percent Purity 99%
Chemical Name or Material Ethylenediaminetetraacetic acid dipotassium salt dihydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-895-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only