Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%, extra pure
CAS: 25102-12-9 | C10H14K2N2O8·2H2O | 404.46 g/mol
Supplier: Thermo Scientific Chemicals 209741000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentSpecifications
| Ethylenediaminetetraacetic acid, dipotassium salt dihydrate | |
| 25102-12-9 | |
| White | |
| Authentic | |
| Plastic bottle | |
| KO2CCH2N(CH2CO2H)CH2CH2N(CH2CO2K)CH2CO2H | |
| 100 g | |
| ethylenediaminetetraacetic acid dipotassium salt dihydrate, dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-bis carboxymethyl amino ethyl azanediyl diacetate dihydrate, edta dipotassium salt dihydrate, edta dipotassium salt, ethylenediaminetetraacetic acid,dipotassium salt dihydrate, potassium ethylenediaminetetraacetate dibasic, dipotassium ethylenediaminetetraacetate dihydrate, ethylenedinitrilo tetraacetic acid dipotassium salt, acmc-209ggx | |
| JFROQFOCFOKDKU-UHFFFAOYSA-L | |
| dipotassium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate | |
| 53486451 | |
| 99% | |
| Fine Crystalline Powder |
| 99% | |
| 272.0°C | |
| 4 to 5 (5% soln.) | |
| 98.5% min. (Complexometry) | |
| C10H14K2N2O8·2H2O | |
| MFCD00150036 | |
| 14, 3517 | |
| (10% in water) Clear colorless | |
| C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[K+].[K+] | |
| 404.46 | |
| 404.46 | |
| Extra Pure |
Chemical Identifiers
| 25102-12-9 | |
| 404.46 | |
| JFROQFOCFOKDKU-UHFFFAOYSA-L | |
| 53486451 | |
| C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[K+].[K+] |
| C10H14K2N2O8·2H2O | |
| MFCD00150036 | |
| ethylenediaminetetraacetic acid dipotassium salt dihydrate, dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-bis carboxymethyl amino ethyl azanediyl diacetate dihydrate, edta dipotassium salt dihydrate, edta dipotassium salt, ethylenediaminetetraacetic acid,dipotassium salt dihydrate, potassium ethylenediaminetetraacetate dibasic, dipotassium ethylenediaminetetraacetate dihydrate, ethylenedinitrilo tetraacetic acid dipotassium salt, acmc-209ggx | |
| dipotassium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
GHS Signal Word: Warning
RUO – Research Use Only