missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ethylenediaminetetraacetic acid dipotassium magnesium salt dihydrate, 97%

Chemical Identifiers

CAS 15708-48-2
Molecular Formula C10H12K2MgN2O8
Molecular Weight (g/mol) 390.71
InChI Key MUEOBEUHFKBRJH-UHFFFAOYSA-J
IUPAC Name magnesium(2+) dipotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
SMILES [Mg++].[K+].[K+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC457941000
View Documents Promotion Details
Thermo Scientific™
457941000
100 g N/A
Only null left
 
AC457945000
View Documents Promotion Details
Thermo Scientific™
457945000
500 g N/A
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

15708-48-2
390.71
magnesium(2+) dipotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
C10H12K2MgN2O8
MUEOBEUHFKBRJH-UHFFFAOYSA-J
[Mg++].[K+].[K+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

15708-48-2
C10H12K2MgN2O8
[Mg++].[K+].[K+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
390.71
Ethylenediaminetetraacetic acid dipotassium magnesium salt dihydrate
96.0 to 104.0 %
MUEOBEUHFKBRJH-UHFFFAOYSA-J
magnesium(2+) dipotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
426.74
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 239-803-8

SDS
Documents

Documents

RUO – Research Use Only