missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic acid diammonium copper salt Solution, volumetric, 0.025 M EDTA-Cu(NH4)2, MilliporeSigma™ Supelco™
Visit our Sensor Applications portal to learn more.
$154.10 - $426.88
Chemical Identifiers
| MDL Number | MFCD01863036 |
|---|---|
| Synonym | Copper diammonium EDTA; Diammonium copper ethylenediaminetetraacetate solution |
Chemical Identifiers
| MFCD01863036 | |
| Copper diammonium EDTA; Diammonium copper ethylenediaminetetraacetate solution |
Specifications
| 105512-06-9 | |
| MFCD01863036 | |
| Copper diammonium EDTA; Diammonium copper ethylenediaminetetraacetate solution | |
| 0.025 M EDTA-Cu(NH4)2 |
| C10H20 CuN4O8 | |
| 250 mL | |
| 387.83 |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label