missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid, Di Na Salt Dihydr. (Crystalline Powd./Electrophor.), Fisher BioReagents™
Description
For minimization of metal ion impurities in reaction buffers
The disodium salt of EDTA is a chelating agent for metal ions and is used extensively in molecular biology to minimize metal ion impurities in reaction buffers.
Specifications
Specifications
| Decomposition Information | >252°C |
| Melting Point | 252°C |
| Color | White |
| pH | 4 to 6 |
| Assay Percent Range | ≥99.0 % |
| Packaging | Poly Bottle |
| Linear Formula | (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O |
| Quantity | 1 kg |
| Merck Index | 15, 3565 |
| Solubility Information | 0.005% max. Insoluble Matter |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:1 Instability:0 |
| Recommended Storage | RT |