missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Cadmium Disodium Salt Hydrate, TCI America™

Catalog No. E008725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E008725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E008725G Supplier TCI America Supplier No. E008725G

Chemical Identifiers

CAS 35803-35-1
Molecular Formula C10H16CdN2Na2O9
Molecular Weight (g/mol) 466.637
MDL Number MFCD00054447
InChI Key AZRABGJSTJUALZ-UHFFFAOYSA-L
Synonym Cadmium Disodium Ethylenediaminetetraacetate
PubChem CID 91886326
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;cadmium;hydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.[Na+].[Na+].[Cd]

Specifications

Color White
Quantity 25 g
UN Number 2570
Formula Weight 446.60
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Cadmium Disodium Salt Hydrate