missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine Anhydrous 98.0+%, TCI America™

Catalog No. p-7128826
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E007725ML 25 mL
E0077500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E007725ML Supplier TCI America Supplier No. E007725ML

Chemical Identifiers

CAS 107-15-3
Molecular Formula C2H8N2
Molecular Weight (g/mol) 60.10
MDL Number MFCD00008204
InChI Key PIICEJLVQHRZGT-UHFFFAOYSA-N
Synonym ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
PubChem CID 3301
ChEBI CHEBI:30347
IUPAC Name ethane-1,2-diamine
SMILES NCCN

Specifications

Color Colorless
pH 11.9 (25% H2O soln. at 25°C)
Boiling Point 116°C
Quantity 25 mL
UN Number 1604
Formula Weight 60.10
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethylenediamine Anhydrous
EINECSNumber (2)-0150
RTECSNumber KH8575000
TSCA Yes